Bethanechol chloride 98%
Catalog # J65919.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
:196.675 g/mol
Merck Index:14,1185
Melting Pt:210…229 °C
EINECS:209-686-8
:GHS07
CAS Number:590-63-6
Formula:C₇H₁₇ClN₂O₂
MDL Number:MFCD00055224