Neohesperidin dihydrochalcone hydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20702-77-6
:MFCD00017711
Formula:C₂₈H₃₆O₁₅.xH₂O
MDL Number:MFCD03840557
:612.58 g/mol (anhy)
Merck Index:14,6452
Storage Temperature:Ambient
EINECS:243-978-6
Related Information
Frequently Bought Together