Order Entry
Italy
ContactUsLinkComponent
Stains-all 95%
Stains-all 95%
Catalog # J62630.03
CAS Number:  
Stains-all 95%
Catalog # J62630.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:7423-31-6
 :
Formula:C₃₀H₂₇BrN₂S₂
MDL Number:MFCD00078788
 :559.584 g/mol
EINECS:231-047-7
 :GHS07