SSC (20X), RNAse free
SSC solution is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0,1X to 20X depending on the application. It is used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts.
Contains 0,3 M sodium citrate, 3,0 M sodium chloride, pH 7,0.
- MDL No.:MFCD00132371
- Physical form:Liquid
Frequently Bought Together