Order Entry
Italy
ContactUsLinkComponent
4-Chloro-2-(trifluoromethyl)pyridine 98%
Toxic
Irritant
4-Chloro-2-(trifluoromethyl)pyridine 98%
Catalog # H64833.06
CAS Number:  
Toxic
Irritant
4-Chloro-2-(trifluoromethyl)pyridine 98%
Catalog # H64833.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

 :
 :181.542 g/mol
ADR:6.1,III
 :GHS06
 :GHS07
Storage Temperature:Ambient
CAS Number:131748-14-6
Formula:C₆H₃ClF₃N
MDL Number:MFCD07368047