Order Entry
Italy
Orders LinkContactUsLinkComponent
5-Bromo-2-hydroxy-4-picoline 97%
5-Bromo-2-hydroxy-4-picoline 97%
5-Bromo-2-hydroxy-4-picoline 97%
5-Bromo-2-hydroxy-4-picoline 97%
Purity: 97% CAS No.: 164513-38-6 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Physical Form: Solid Appearance: Off-white Melting Point: 198-202° Beilstein Reference: 6798531 MDL No.: MFCD03427664
 :  5-Bromo-2-hydroxy-4-methylpyridine
  •  :164513-38-6
  •  :C₆H₆BrNO
  •  :MFCD03427664
  •  :188.02 g/mol
  •  :198…202 °C
  •  :Ambient
Product Family Options
  • Pk
  • 5 g
    1 * 5 g
    1 * 5 g - 55,40 €
    55,40 €
    1 * 5 g
       55,40 €
  • 25 g
    1 * 25 g
    1 * 25 g - 192,00 €
    192,00 €
    1 * 25 g
       192,00 €