
Methyl-4-methoxypicolinate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29681-43-4
:
Formula:C₈H₉NO₃
MDL Number:MFCD00956223
:167.16 g/mol
Melting Pt:47…50 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together