
Methyl-2-amino-4,5-difluorobenzoate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207346-42-7
:
Formula:C₈H₇F₂NO₂
MDL Number:MFCD09264511
:187.15 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together