
4-Amino-5-bromopyrimidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1439-10-7
:
Formula:C₄H₄BrN₃
MDL Number:MFCD00233962
Melting Pt:207…211 °C
:174.00 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together