
5-(Trifluoromethyl)anthranilic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:83265-53-6
:
Formula:C₈H₆F₃NO₂
MDL Number:MFCD01569537
:205.14 g/mol
Melting Pt:141…146 °C (dec.)
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together