
1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1141878-45-6
:
Formula:C₉H₁₂BF₃N₂O₃
MDL Number:MFCD17214248
:264.01 g/mol
:GHS07
Frequently Bought Together