Methyl-2-bromothiazole-4-carboxylate 96%
Catalog # H63541.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
:
:222.06 g/mol
Storage Temperature:Ambient
CAS Number:170235-26-4
Formula:C₅H₄BrNO₂S
MDL Number:MFCD06659908