

4,5-Difluoro-2-nitrotoluene 98%
Catalog # H63030.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
CAS Number:127371-50-0
:
Formula:C₇H₅F₂NO₂
MDL Number:MFCD04973772
ADR:6.1,III
:173.12 g/mol
UN:2810
:GHS06
:GHS07