H-ORN(Z)-OH 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3304-51-6
:
Formula:C₁₃H₁₈N₂O₄
MDL Number:MFCD00037220
:266.297 g/mol
Melting Pt:248…252 °C
Storage Temperature:Freezer
EINECS:221-980-8
Related Information
Frequently Bought Together