2,6-Difluoro-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Phenol 96%
Catalog # H62042.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 mg
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3,5-Difluoro-4-hydroxybenzeneboronic acid pinacol ester
CAS Number:1029439-83-5
:
Formula:C₁₂H₁₅BF₂O₃
:256.056 g/mol
Storage Temperature:Ambient
MDL Number:MFCD16996286