
3-Bromo-5-methylaniline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74586-53-1
:
Formula:C7H8BrN
MDL Number:MFCD11845553
:186.05 g/mol
Storage Temperature:Ambient
EINECS:277-932-1
ADR:6.1,III
:GHS07
Frequently Bought Together