

4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:82925-02-8
:
Formula:C₁₉H₂₄N₂O₂
MDL Number:MFCD04974539
:312.41 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together