Boc-3,4-dichloro-L-phenylalanine 95%
Catalog # H52053.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:80741-39-5
:
Formula:C₁₄H₁₇Cl₂NO₄
MDL Number:MFCD00273436
:334.2 g/mol
Storage Temperature:Ambient