N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:114873-01-7
:
Formula:C₁₄H₁₈FNO₄
MDL Number:MFCD00672522
:283.30 g/mol
Melting Pt:75…80 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together