Order Entry
Italy
ContactUsLinkComponent
3-Methylpyridine-2-thiocarboxamide 97%
Toxic
Irritant
3-Methylpyridine-2-thiocarboxamide 97%
Catalog # H51794.03
CAS Number:  
Toxic
Irritant
3-Methylpyridine-2-thiocarboxamide 97%
Catalog # H51794.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.21 g/mol
Melting Pt:117…122 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient
CAS Number:334017-95-7
Formula:C₇H₈N₂S
MDL Number:MFCD12026307