N-Boc-1,3-propanediamine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75178-96-0
:
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD00210021
:174.244 g/mol
Melting Pt:∼ 22 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05
Related Information
Frequently Bought Together