
Thieno[3,2-b]pyridin-7-ol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107818-20-2
:
Formula:C₇H₅NOS
MDL Number:MFCD01863636
:151.183 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together