Order Entry
Italy
ContactUsLinkComponent
6-Bromo-2-chloro-1,8-naphthyridine 96%
Irritant
6-Bromo-2-chloro-1,8-naphthyridine 96%
Catalog # H50043.03
CAS Number:  
Irritant
6-Bromo-2-chloro-1,8-naphthyridine 96%
Catalog # H50043.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:902837-40-5
 :
Formula:C₈H₄BrClN₂
MDL Number:MFCD07781152
 :243.49 g/mol
 :GHS07
Storage Temperature:Ambient