
Lithium-4,5-dicyano-2-(trifluoromethyl)imidazole 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:C₆F₃LiN₄
MDL Number:MFCD18827480
:192.03 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together