Order Entry
Italy
ContactUsLinkComponent
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
Irritant
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
Catalog # H33292.03
CAS Number:  
Irritant
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
Catalog # H33292.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.59 g/mol
Melting Pt:108…110 °C
 :GHS07
Storage Temperature:Ambient
Formula:C₈H₆ClNO₂
MDL Number:MFCD20040113