Di-2-thienyl sulphide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3988-99-6
:
Formula:C₈H₆S₃
MDL Number:MFCD00052047
Boiling Pt:102…104 °C (0.9 mmHg)
Density:1.311
Flash Pt:>100 °C (212 °F)
:198.32 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together