
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde ≥98%
Catalog # H32602.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:53104-95-3
:
Formula:C₈H₅F₃O₃
MDL Number:MFCD04973780
:206.12 g/mol
Melting Pt:76…80 °C
:GHS07
Storage Temperature:Ambient