
Ethyl-2-bromo-4,4,4-trifluorobutyrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:367-33-9
:
Formula:C₆H₈BrF₃O₂
MDL Number:MFCD10687235
:249.03 g/mol
Boiling Pt:67 °C (18 mmHg)
Storage Temperature:Ambient
ADR:8,II
:GHS05
Frequently Bought Together