

2,2-Difluorocyclopropanemethanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:509072-57-5
:
Formula:C₄H₆F₂O
MDL Number:MFCD06797748
:108.09 g/mol
Boiling Pt:129 °C
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together