
Ethyl-2-(trifluoromethyl)-4-methyl-5-pyrimidine carboxylate 97%
Catalog # H32065.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:306960-67-8
:
Formula:C₉H₉F₃N₂O₂
MDL Number:MFCD07367386
:234.18 g/mol
Boiling Pt:54 °C (0.5 mmHg)
:GHS07
Storage Temperature:Ambient