Order Entry
Italy
ContactUsLinkComponent
 
Diethyl-4,4-difluoroheptanedioate 97%
Catalog # H31544.03
CAS Number:  
Diethyl-4,4-difluoroheptanedioate 97%
Catalog # H31544.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :252.26 g/mol
Boiling Pt:108 °C (3 mmHg)
Storage Temperature:Ambient
CAS Number:22515-16-8
Formula:C₁₁H₁₈F₂O₄
MDL Number:MFCD08146634