Indium ≥99.99% (metals basis), rods 2.54 mm (0.10 in) Ø
Catalog # 43249.BV
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:30 cm
- Seq:7765
Specifications
About this item
:
:114.82 g/mol
Merck Index:13,04971
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:156 °C
Density:7.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-180-0
CAS Number:7440-74-6
Formula:In
MDL Number:MFCD00134048