Order Entry
Italy
ContactUsLinkComponent
Ethyl-2-aminothiazole-4-carboxylate 98%
Irritant
Ethyl-2-aminothiazole-4-carboxylate 98%
Catalog # H30634.09
CAS Number:  
Irritant
Ethyl-2-aminothiazole-4-carboxylate 98%
Catalog # H30634.09
CAS Number:  

Specifications

  • Pk:
    10 g

Specifications

About this item

 :
 :172.21 g/mol
Melting Pt:175…178 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:5398-36-7
Formula:C₆H₈N₂O₂S
MDL Number:MFCD00619079