
Ethyl-α-4-dibromophenylacetate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77143-76-1
:
Formula:C₁₀H₁₀Br₂O₂
MDL Number:MFCD09953497
:321.99 g/mol
Boiling Pt:140 °C (5 mmHg)
Density:1.642
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
ADR:8,II
:GHS05
Frequently Bought Together