
1,1'-(Azodicarbonyl)dipiperidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10465-81-3
:
Formula:C₁₂H₂₀N₄O₂
MDL Number:MFCD00010111
:252.32 g/mol
Melting Pt:133…136 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together