Order Entry
Italy
ContactUsLinkComponent
4,5-Dimethylthiophene-2-carboxaldehyde 97%
Irritant
4,5-Dimethylthiophene-2-carboxaldehyde 97%
Catalog # H27712.03
CAS Number:  
Irritant
4,5-Dimethylthiophene-2-carboxaldehyde 97%
Catalog # H27712.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:5928-48-3
 :
Formula:C₇H₈OS
 :140.2 g/mol
Boiling Pt:82…85 °C
Density:1.179
Flash Pt:>110 °C (230 °F)
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD02093897