
Diethyl allyl(3-phenyl-2-propynyl)malonate 96%
Codice catalogo H27702.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:143633-91-4
:
Formula:C₁₉H₂₂O₄
MDL Number:MFCD09953493
:314.38 g/mol
:GHS07
Storage Temperature:Ambient