1-O-Methyl-2-deoxy-D-ribose 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60134-26-1
:
Formula:C₆H₁₂O₄
MDL Number:MFCD00134161
:148.17 g/mol
Density:1.224
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together