
4-Bromomethyl-1,2-dihydroquinoline-2-one
Catalog # B25305.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
CAS Number:4876-10-2
:
Formula:C₁₀H₈BrNO
:238.08 g/mol
Melting Pt:∼ 254 °C (dec.)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
MDL Number:MFCD03426152