Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Formaldoxime trimer hydrochloride
Formaldoxime trimer hydrochloride 95%
Formaldoxime trimer hydrochloride 95%
B25063.09
:
Thermo Fisher Scientific
:
Formaldoxime trimer hydrochloride 95%
B25063.09
:
Thermo Fisher Scientific
:
Pk:
10 g
Seq:
3236
:
6286-29-9
:
:
C₃H₉N₃O₃·HCl
:
MFCD00012778
:
171.57 g/mol
:
∼ 128 °C (dec.)
:
222-440-4
:
6.1,II
:
GHS06
:
GHS07
:
Ambient