2,2'-Furil dioxime mixture of isomers 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:522-27-0
:
Formula:C₁₀H₈N₂O₄
MDL Number:MFCD00075315
:220.19 g/mol
Merck Index:14,4306
Melting Pt:∼ 165 °C
Storage Temperature:Ambient
EINECS:208-326-7
Related Information
Certificates
Search by Lot Numbermsds_ALFAB23972
Scheda di sicurezza
Organic Building Blocks and Intermediates
Note applicative
Frequently Bought Together