1,1-Diphenyl-2-propyn-1-ol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3923-52-2
:
Formula:C₁₅H₁₂O
MDL Number:MFCD00041570
:208.26 g/mol
Boiling Pt:179…180 °C (18 mmHg)
Melting Pt:44…48 °C
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together