2-Methyl-4-octanone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7492-38-8
:
Formula:C₉H₁₈O
MDL Number:MFCD00048808
:142.24 g/mol
Boiling Pt:180 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together