
3-Bromo-2-(bromomethyl)propionic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:41459-42-1
:
Formula:C₄H₆Br₂O₂
MDL Number:MFCD00010643
:245.90 g/mol
Melting Pt:98…101 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05
Related Information
Frequently Bought Together