Farnesylacetone mixture of isomers 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:762-29-8
:
Formula:C₁₈H₃₀O
MDL Number:MFCD00036517
:262.44 g/mol
Density:0.880
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:212-097-9
Related Information
Frequently Bought Together