
(E/Z)-Geranylacetone mixture E/Z 97% (Z)-isomer (nerylacetone) ∼ 45%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:689-67-8
:
Formula:(CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(O)CH₃
MDL Number:MFCD00008910
:194.32 g/mol
Boiling Pt:250 °C
Density:0.87
Flash Pt:110 °C (230 °F)
EINECS:211-711-2
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together