Acid red 52
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 3520-42-1 Molecular Formula: C27H29N2NaO7S2 Formula Weight: 580.66 Beilstein Reference: 3886008 MDL No.: MFCD00010180
Synonyms: Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200
- CAS Number:3520-42-1
- Formula:C₂₇H₃₀N₂O₇S₂
- MDL Number:MFCD00010180
- Molecular Weight:580.65 g/mol
- Storage Temperature:Ambient
- EINECS:222-529-8
2 Options Available Below
Product Details & Documents
CAS No.: 3520-42-1 Molecular Formula: C27H29N2NaO7S2 Formula Weight: 580.66 Beilstein Reference: 3886008 MDL No.: MFCD00010180
Synonyms: Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200
- CAS Number:3520-42-1
- Formula:C₂₇H₃₀N₂O₇S₂
- MDL Number:MFCD00010180
- Molecular Weight:580.65 g/mol
- Storage Temperature:Ambient
- EINECS:222-529-8
Product Family Options
Product Information
- PkAvailabilityPrice



