Methyl-1-methyl-1H-pyrrole-2-carboxylate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37619-24-2
:
Formula:C₇H₉NO₂
MDL Number:MFCD00052747
:139.16 g/mol
Boiling Pt:96…98 °C (15 mmHg)
Storage Temperature:Ambient
Related Information
Frequently Bought Together