1-Benzylpiperidin-3-one hydrochloride hydrate, hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50606-58-1
Formula:C₁₂H₁₅NO.HCl.xH₂O
MDL Number:MFCD00150096
:225.72 g/mol (anhy)
Melting Pt:105…107 °C
Storage Temperature:Refrigerator
EINECS:256-646-0
Related Information
Frequently Bought Together