Order Entry
Italy
ContactUsLinkComponent
Methylcobalamin hydrate 97% (by HPLC)
Methylcobalamin hydrate 97% (by HPLC)
Certificates
Methylcobalamin hydrate 97% (by HPLC)
Certificates

Specification Test Results

  • Appearance (Color):
    Red to Dark red to brown
  • Form:
    Powder or crystals
  • Assay (HPLC):
    ≥96.0%
  • Water Content (Karl Fischer Titration):
    <12.0% (<10 waters of hydration)

Specification Test Results

Related Information

Frequently Bought Together